Assay Details
Gene Symbol: Emc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495108.2 | XP_003495156.1 | 4 - 5 | 590 | 56 | |
GenBank mRNA | JI884097.1 | - | 4 - 5 | 575 | 56 | |
JP049933.1 | - | 4 - 5 | 575 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
