Assay Details
| Gene Symbol: Pms2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496274.2 | XP_003496322.1 | 2 - 3 | 231 | 73 | |
| XM_007648692.1 | XP_007646882.1 | 2 - 3 | 307 | 73 | ||
More Information
Set Membership: |
|

Pms2
Genomic Map