Assay Details
| Gene Symbol: Mtfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496549.2 | XP_003496597.2 | 5 - 6 | 704 | 63 | |
| GenBank mRNA | JI876843.1 | - | 4 - 5 | 560 | 63 | |
More Information
Set Membership: |
|

Mtfr2
Genomic Map