Assay Details
| Gene Symbol: Ostm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496980.2 | XP_003497028.1 | 1 - 2 | 468 | 61 | |
| GenBank mRNA | JP037172.1 | - | 1 - 2 | 243 | 61 | |
More Information
Set Membership: |
|

Ostm1
Genomic Map