Assay Details
| Gene Symbol: Tsen15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500853.2 | XP_003500901.2 | 2 - 3 | 371 | 83 | |
| GenBank mRNA | JI874864.1 | - | 371 | 83 | ||
More Information
Set Membership: |
|

Tsen15
Genomic Map