Assay Details
| Gene Symbol: Acot13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502191.2 | XP_003502239.1 | 2 - 3 | 426 | 92 | |
| GenBank mRNA | JI870939.1 | - | 2 - 3 | 411 | 92 | |
| JP047507.1 | - | 2 - 3 | 412 | 92 | ||
More Information
Set Membership: |
|

Acot13
Genomic Map