Assay Details
| Gene Symbol: LOC100756672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510133.2 | XP_003510181.1 | 12 - 13 | 1966 | 90 | |
| GenBank mRNA | JI865661.1 | - | 4 - 5 | 591 | 90 | |
| JP059944.1 | - | 4 - 5 | 591 | 90 | ||
More Information
Set Membership: |
|

LOC100756672
Genomic Map