Assay Details
| Gene Symbol: LOC100756672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510133.2 | XP_003510181.1 | 13 - 14 | 2073 | 83 | |
| GenBank mRNA | JI865661.1 | - | 5 - 6 | 698 | 83 | |
| JP059944.1 | - | 5 - 6 | 698 | 83 | ||
More Information
Set Membership: |
|

LOC100756672
Genomic Map