Assay Details
| Gene Symbol: LOC100756672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510133.2 | XP_003510181.1 | 2 - 3 | 806 | 79 | |
| GenBank mRNA | JI869365.1 | - | 2 - 3 | 275 | 79 | |
| JP049717.1 | - | 2 - 3 | 275 | 79 | ||
More Information
Set Membership: |
|

LOC100756672
Genomic Map