Assay Details
| Gene Symbol: Psma5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510965.2 | XP_003511013.1 | 5 - 6 | 525 | 68 | |
| GenBank mRNA | JI866973.1 | - | 5 - 6 | 532 | 68 | |
More Information
Set Membership: |
|

Psma5
Genomic Map