Assay Details
| Gene Symbol: Ormdl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607960.1 | XP_007606150.1 | 2 - 3 | 455 | 61 | |
| XM_007607961.1 | XP_007606151.1 | 2 - 3 | 450 | 61 | ||
More Information
Set Membership: |
|

Ormdl3
Genomic Map