Assay Details
| Gene Symbol: Fam150a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647988.1 | XP_007646178.1 | 5 - 6 | 465 | 130 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam150a| Gene Symbol: Fam150a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647988.1 | XP_007646178.1 | 5 - 6 | 465 | 130 | |
Set Membership: |
|