Assay Details
Gene Symbol: Barhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653837.1 | XP_007652027.1 | 4 - 5 | 1101 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
