Assay Details
| Gene Symbol: Fhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502239.2 | XP_003502287.2 | 2 - 3 | 249 | 63 | |
| XM_007642852.1 | XP_007641042.1 | 2 - 3 | 308 | 63 | ||
| XM_007642853.1 | XP_007641043.1 | 1 - 2 | 193 | 63 | ||
| GenBank mRNA | JI896430.1 | - | 1 - 2 | 201 | 63 | |
More Information
Set Membership: |
|

Fhl3
Genomic Map