Assay Details
| Gene Symbol: Adnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502474.2 | XP_003502522.1 | 3 - 4 | 584 | 69 | |
| XM_007643098.1 | XP_007641288.1 | 3 - 4 | 722 | 69 | ||
More Information
Set Membership: |
|

Adnp
Genomic Map