Assay Details
| Gene Symbol: Rab5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506195.2 | XP_003506243.1 | 3 - 4 | 512 | 78 | |
| XM_007646702.1 | XP_007644892.1 | 4 - 5 | 573 | 78 | ||
| XM_007646703.1 | XP_007644893.1 | 3 - 4 | 535 | 78 | ||
| XM_007646704.1 | XP_007644894.1 | 3 - 4 | 564 | 78 | ||
| GenBank mRNA | JI867395.1 | - | 3 - 4 | 507 | 78 | |
| JP039010.1 | - | 3 - 4 | 506 | 78 | ||
More Information
Set Membership: |
|

Rab5b
Genomic Map