Assay Details
| Gene Symbol: Tomm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507442.2 | XP_003507490.1 | 2 - 3 | 237 | 72 | |
| GenBank mRNA | JI888833.1 | - | 2 - 3 | 238 | 72 | |
More Information
Set Membership: |
|

Tomm7
Genomic Map