Assay Details
| Gene Symbol: Rrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509476.2 | XP_003509524.1 | 7 - 8 | 891 | 69 | |
| GenBank mRNA | JI889589.1 | - | 7 - 8 | 873 | 69 | |
| JP043835.1 | - | 7 - 8 | 873 | 69 | ||
More Information
Set Membership: |
|

Rrm2
Genomic Map