Assay Details
| Gene Symbol: LOC100755585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648086.1 | XP_007646276.1 | 17 - 18 | 2080 | 76 | |
| XM_007648087.1 | XP_007646277.1 | 16 - 17 | 1983 | 76 | ||
| GenBank mRNA | JI882223.1 | - | 5 - 6 | 936 | 76 | |
| JP051300.1 | - | 5 - 6 | 933 | 76 | ||
More Information
Set Membership: |
|

LOC100755585
Genomic Map