Assay Details
| Gene Symbol: LOC100755585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648086.1 | XP_007646276.1 | 18 - 19 | 2239 | 76 | |
| XM_007648087.1 | XP_007646277.1 | 17 - 18 | 2142 | 76 | ||
| GenBank mRNA | JI882223.1 | - | 6 - 7 | 1095 | 76 | |
| JP051300.1 | - | 6 - 7 | 1092 | 76 | ||
More Information
Set Membership: |
|

LOC100755585
Genomic Map