Assay Details
| Gene Symbol: LOC100755585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648086.1 | XP_007646276.1 | 23 - 24 | 2845 | 87 | |
| XM_007648087.1 | XP_007646277.1 | 22 - 23 | 2748 | 87 | ||
| GenBank mRNA | JI861946.1 | - | 2 - 3 | 187 | 87 | |
| JP059464.1 | - | 2 - 3 | 187 | 87 | ||
More Information
Set Membership: |
|

LOC100755585
Genomic Map