Assay Details
| Gene Symbol: LOC100755585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648086.1 | XP_007646276.1 | 29 - 30 | 3710 | 69 | |
| XM_007648088.1 | XP_007646278.1 | 7 - 8 | 757 | 69 | ||
More Information
Set Membership: |
|

LOC100755585
Genomic Map