Assay Details
| Gene Symbol: LOC100755585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648086.1 | XP_007646276.1 | 4 - 5 | 450 | 83 | |
| XM_007648087.1 | XP_007646277.1 | 4 - 5 | 449 | 83 | ||
| GenBank mRNA | JI882222.1 | - | 4 - 5 | 445 | 83 | |
| JP051299.1 | - | 4 - 5 | 445 | 83 | ||
More Information
Set Membership: |
|

LOC100755585
Genomic Map