Assay Details
| Gene Symbol: LOC100770827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644528.1 | XP_007642718.1 | 11 - 12 | 2005 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100770827| Gene Symbol: LOC100770827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644528.1 | XP_007642718.1 | 11 - 12 | 2005 | 101 | |
Set Membership: |
|