Assay Details
Gene Symbol: Ppap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495751.2 | XP_003495799.1 | 2 - 3 | 543 | 98 | |
GenBank mRNA | JI869948.1 | - | 2 - 3 | 690 | 98 | |
JP063324.1 | - | 2 - 3 | 690 | 98 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
