Assay Details
| Gene Symbol: Crabp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497903.2 | XP_003497951.1 | 1 - 2 | 65 | 69 | |
| XM_007619309.1 | XP_007617499.1 | 1 - 2 | 65 | 69 | ||
More Information
Set Membership: |
|

Crabp1
Genomic Map