Assay Details
| Gene Symbol: Slc16a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498966.2 | XP_003499014.1 | 3 - 4 | 358 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc16a12| Gene Symbol: Slc16a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498966.2 | XP_003499014.1 | 3 - 4 | 358 | 72 | |
Set Membership: |
|