Assay Details
| Gene Symbol: Cklf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499679.2 | XP_003499727.1 | 2 - 3 | 387 | 103 | |
| GenBank mRNA | JI863545.1 | - | 2 - 3 | 357 | 103 | |
| JP042394.1 | - | 2 - 3 | 354 | 103 | ||
More Information
Set Membership: |
|

Cklf
Genomic Map