Assay Details
| Gene Symbol: Cklf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499679.2 | XP_003499727.1 | 3 - 4 | 490 | 87 | |
| GenBank mRNA | JI863545.1 | - | 3 - 4 | 460 | 87 | |
| JP042394.1 | - | 3 - 4 | 457 | 87 | ||
More Information
Set Membership: |
|

Cklf
Genomic Map