Assay Details
| Gene Symbol: Icoslg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500605.1 | XP_003500653.1 | 7 - 8 | 1654 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Icoslg| Gene Symbol: Icoslg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500605.1 | XP_003500653.1 | 7 - 8 | 1654 | 60 | |
Set Membership: |
|