Assay Details
| Gene Symbol: Rab28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507955.2 | XP_003508003.1 | 1 - 2 | 261 | 78 | |
| XM_007648557.1 | XP_007646747.1 | 1 - 2 | 261 | 78 | ||
| XM_007648558.1 | XP_007646748.1 | 1 - 2 | 261 | 78 | ||
| XM_007648559.1 | XP_007646749.1 | 1 - 2 | 261 | 78 | ||
| GenBank mRNA | JI878456.1 | - | 1 - 2 | 176 | 78 | |
| JP056359.1 | - | 1 - 2 | 176 | 78 | ||
More Information
Set Membership: |
|

Rab28
Genomic Map