Assay Details
| Gene Symbol: Fam195b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496937.2 | XP_003496985.1 | 3 - 4 | 3192 | 78 | |
| GenBank mRNA | JI875029.1 | - | 1 - 2 | 111 | 78 | |
| JP047519.1 | - | 1 - 2 | 111 | 78 | ||
More Information
Set Membership: |
|

Fam195b
Genomic Map