Assay Details
| Gene Symbol: LOC100763079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498198.2 | XP_003498246.1 | 1 - 2 | 37 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100763079| Gene Symbol: LOC100763079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498198.2 | XP_003498246.1 | 1 - 2 | 37 | 61 | |
Set Membership: |
|