Assay Details
| Gene Symbol: Eef1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503130.2 | XP_003503178.1 | 6 - 7 | 1166 | 54 | |
| XM_007643761.1 | XP_007641951.1 | 5 - 6 | 1112 | 54 | ||
More Information
Set Membership: |
|

Eef1a2
Genomic Map