Assay Details
| Gene Symbol: Sap30bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506815.2 | XP_003506863.1 | 5 - 6 | 439 | 72 | |
| XM_007647357.1 | XP_007645547.1 | 5 - 6 | 442 | 72 | ||
| GenBank mRNA | JI865311.1 | - | 2 - 3 | 132 | 72 | |
| JP046650.1 | - | 2 - 3 | 134 | 72 | ||
More Information
Set Membership: |
|

Sap30bp
Genomic Map