Assay Details
| Gene Symbol: Psmd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007631458.1 | XP_007629648.1 | 6 - 7 | 697 | 60 | |
| XM_007631465.1 | XP_007629655.1 | 6 - 7 | 697 | 60 | ||
| GenBank mRNA | JI874891.1 | - | 6 - 7 | 702 | 60 | |
More Information
Set Membership: |
|

Psmd4
Genomic Map