Assay Details
| Gene Symbol: LOC100758857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645488.1 | XP_007643678.1 | 2 - 3 | 1697 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100758857| Gene Symbol: LOC100758857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645488.1 | XP_007643678.1 | 2 - 3 | 1697 | 80 | |
Set Membership: |
|