Assay Details
| Gene Symbol: Nkiras1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498209.2 | XP_003498257.1 | 3 - 4 | 555 | 99 | |
| GenBank mRNA | JI869221.1 | - | 2 - 3 | 277 | 99 | |
| JP058619.1 | - | 2 - 3 | 277 | 99 | ||
More Information
Set Membership: |
|

Nkiras1
Genomic Map