Assay Details
| Gene Symbol: Znf385b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501027.2 | XP_003501075.2 | 8 - 9 | 1335 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znf385b| Gene Symbol: Znf385b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501027.2 | XP_003501075.2 | 8 - 9 | 1335 | 65 | |
Set Membership: |
|