Assay Details
| Gene Symbol: Stam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503377.2 | XP_003503425.1 | 13 - 14 | 1582 | 63 | |
| GenBank mRNA | JI887223.1 | - | 13 - 14 | 1582 | 63 | |
| JP048155.1 | - | 13 - 14 | 1582 | 63 | ||
More Information
Set Membership: |
|

Stam
Genomic Map