Assay Details
| Gene Symbol: Fbxl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503735.1 | XP_003503783.1 | 2 - 3 | 483 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxl21| Gene Symbol: Fbxl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503735.1 | XP_003503783.1 | 2 - 3 | 483 | 68 | |
Set Membership: |
|