Assay Details
| Gene Symbol: Spty2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509345.2 | XP_003509393.1 | 5 - 6 | 2156 | 70 | |
| XM_007649930.1 | XP_007648120.1 | 6 - 7 | 2099 | 70 | ||
| GenBank mRNA | JI875009.1 | - | 3 - 4 | 608 | 70 | |
| JP065245.1 | - | 3 - 4 | 608 | 70 | ||
More Information
Set Membership: |
|

Spty2d1
Genomic Map