Assay Details
| Gene Symbol: Agpat9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514868.2 | XP_003514916.1 | 1 - 2 | 531 | 74 | |
| XM_007654908.1 | XP_007653098.1 | 2 - 3 | 469 | 74 | ||
| XM_007654909.1 | XP_007653099.1 | 2 - 3 | 470 | 74 | ||
More Information
Set Membership: |
|

Agpat9
Genomic Map