Assay Details
| Gene Symbol: Hadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650262.1 | XP_007648452.1 | 3 | 670 | 76 | |
| GenBank mRNA | JP038226.1 | - | 3 | 474 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hadh| Gene Symbol: Hadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650262.1 | XP_007648452.1 | 3 | 670 | 76 | |
| GenBank mRNA | JP038226.1 | - | 3 | 474 | 76 | |
Set Membership: |
|