Assay Details
| Gene Symbol: Arhgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497298.2 | XP_003497346.1 | 5 - 6 | 551 | 58 | |
| XM_007610085.1 | XP_007608275.1 | 5 - 6 | 517 | 58 | ||
| XM_007610103.1 | XP_007608293.1 | 5 - 6 | 515 | 58 | ||
| GenBank mRNA | JI884817.1 | - | 5 - 6 | 552 | 58 | |
| JP062864.1 | - | 5 - 6 | 552 | 58 | ||
More Information
Set Membership: |
|

Arhgap1
Genomic Map