Assay Details
| Gene Symbol: Wfdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504582.1 | XP_003504630.1 | 2 - 3 | 218 | 82 | |
| XM_007645117.1 | XP_007643307.1 | 2 - 3 | 221 | 82 | ||
More Information
Set Membership: |
|

Wfdc2
Genomic Map