Assay Details
| Gene Symbol: Leprot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508137.2 | XP_003508185.1 | 2 - 3 | 162 | 64 | |
| XM_007648761.1 | XP_007646951.1 | 2 - 3 | 354 | 64 | ||
| GenBank mRNA | JI885474.1 | - | 2 - 3 | 157 | 64 | |
| JP064232.1 | - | 2 - 3 | 157 | 64 | ||
More Information
Set Membership: |
|

Leprot
Genomic Map