Assay Details
| Gene Symbol: Leprot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508137.2 | XP_003508185.1 | 3 - 4 | 342 | 120 | |
| XM_007648761.1 | XP_007646951.1 | 3 - 4 | 534 | 120 | ||
| GenBank mRNA | JI885474.1 | - | 3 - 4 | 337 | 120 | |
| JP064232.1 | - | 3 - 4 | 337 | 120 | ||
More Information
Set Membership: |
|

Leprot
Genomic Map