Assay Details
| Gene Symbol: Fam84b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515680.1 | XP_003515728.1 | 1 | 719 | 87 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 86 and 100  3' Most  Detect Genomic DNA  Recommended Assay  Both primers and probe map within a single exon | 
 Back To Top
 Back To Top
 Fam84b
Fam84b Genomic Map
Genomic Map Back To Top
 Back To Top