Assay Details
| Gene Symbol: LOC100760380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607801.1 | XP_007605991.1 | 10 - 11 | 1744 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100760380| Gene Symbol: LOC100760380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607801.1 | XP_007605991.1 | 10 - 11 | 1744 | 120 | |
Set Membership: |
|